Nuclear Green™ DCS1
Leverandør: AAT BIOQUEST
17550
AATB17550EA 1800 NOK
AATB17550
Nuclear Green™ DCS1
Assay Cellulær analyse
Our Nuclear Green™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analysing DNA content in dead, fixed or apoptotic cells.
  • Cell-impermeable
  • Useful for multicolour imaging
  • Bright fluorescence
5 mM DMSO solution.
NB: For research use only.