Helixyte™ Green *20X Aqueous PCR Solution*
Leverandør: AAT BIOQUEST
17591
AATB17591EA 3600 NOK
AATB17591
Helixyte™ Green *20X Aqueous PCR Solution*
Assay qPCR analyser
Helixyte™ Gold is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Gold (SYBR® is the trademark of ThermoFisher).
  • Same spectral properties to SYBR® gold
  • High sensitivity
  • Useful for nucleic acid labeling in gels
NB: For research use only.