Helixyte™ Green *10000X Aqueous PCR Solution*
Leverandør: AAT BIOQUEST
17592
AATB17592EA 17325 NOK
AATB17592
Helixyte™ Green *10000X Aqueous PCR Solution*
Assay qPCR analyser
Helixyte™ Green is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Green (SYBR® is the trademark of ThermoFisher).
  • Same Spectral Properties To SYBR® Green
  • High Sensitivity
  • Useful For Nucleic Acid Labeling In Gels
NB: For research use only.