Portelite™ Fluorimetric ssDNA Quantitation Kit
Leverandør: AAT BIOQUEST
17625
AATB17625EA 5000 NOK
AATB17625
Portelite™ Fluorimetric ssDNA Quantitation Kit
Assay Nucleic Acid Assays
The assay detects long oligonucleotides or ssDNA. Nucleotides and short oligo nucleotides of six bases or less do not interfere in the quantitation assay.
  • Optimized for cytocite and qubit fluorometers
  • Any volume from 1 to 20 µl is acceptable
  • Assay range of 1 to 200 ng
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer, and prediluted DNA standards. Helixyte™ Green ssDNA reagent is a sensitive fluorescent nucleic acid probe for quantifying oligonucleotides and single-stranded DNA (ssDNA) in solution.
Forpakning: Plastic vial.
NB: For research use only.