StrandBrite™ Green Fluorimetric RNA Quantitation Kit
Leverandør: AAT BIOQUEST
17656
AATB17656EA 5400 NOK
AATB17656
StrandBrite™ Green Fluorimetric RNA Quantitation Kit
Assay Nucleic Acid Assays
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, reverse transcription PCR, and differential display PCR.
  • Highly sensitive RNA detection
  • Robust protocol
  • Multiple applications
NB: For research use only.