Gelite™ Safe DNA Gel Stain *GelRed Replacement, 10000X in Water*
Leverandør: AAT BIOQUEST
17708
AATB17708EA 1625 NOK
AATB17708
Gelite™ Safe DNA Gel Stain *GelRed Replacement, 10000X in Water*
Farger Elektroforese Farging
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
  • Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
NB: For research use only.