Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in DMSO*
Leverandør: AAT BIOQUEST
17711
AATB17711EA 1625 NOK
AATB17711
Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in DMSO*
Farger Elektroforese Farging
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
  • Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
NB: For research use only.