XFD647 alkyne
Leverandør: AAT BIOQUEST
1897
AATB1897EA 3250 NOK
AATB1897
XFD647 alkyne
XFD647 alkyne
XFD647 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 647 (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only