ReadiUse™ TCA Deproteinization Sample Preparation Kit
Leverandør: AAT BIOQUEST
19501
AATB19501EA 2700 NOK
AATB19501
ReadiUse™ TCA Deproteinization Sample Preparation Kit
Assay Proteinanalyser
The analysis of amino acids, ions, metabolites and other small molecules is frequently hindered by the interference of lipids, protein and enzymes present in biofluids, cell and tissue lysates.
  • Contains all necessary components
  • Useful for preparing small molecules
  • Can be used in small- and large-scale
NB: For research use only.