FluoroQuest™ Fluorescence Quantum Yield Determination Kit *Optimized for Bioconjugates* (1 kit)

Leverandør: AAT BIOQUEST

20
AATB20EA 17325 NOK
AATB20
FluoroQuest™ Fluorescence Quantum Yield Determination Kit *Optimized for Bioconjugates* (1 kit)
Assay Nucleic Acid Assays

When a fluorophore absorbs a photon of light, an energetically excited state is formed.


  • Robust protocol
  • Contains all necessary components


NB: For research use only.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR