Pluronic® F-127 Cell Culture Tested
Leverandør: AAT BIOQUEST
20050
AATB20050EA 1425 NOK
AATB20050
Pluronic® F-127 Cell Culture Tested
Rengjøringsløsninger Detergenter og overflatevasking
Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.
  • Non-toxic to cells
It is soluble at low temperature, but gels at room temperature.