Pluronic® F-127 10% water

Leverandør: AAT BIOQUEST
Warning

Synonymer: PEG-PPG-PEG, Pluronic® F-127

20053
AATB20053EA 1425 NOK
AATB20053
Pluronic® F-127 10% water
Poloxamer 407 (Pluronic® F-127)

Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.


  • Extremely common surfectant
  • Multiple applications
  • Aqueous solution


NB: For research use only.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082049
CAS-nummer: 9003-11-6
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR