Annexin V-Cy5.5 Conjugate
Leverandør: AAT BIOQUEST
20067
AATB20067EA 3600 NOK
AATB20067
Annexin V-Cy5.5 Conjugate
Assay Cellulær analyse
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
  • High stability
  • In-house quality control
  • Multiple research applications
NB: For research use only.