Cell Meter™ Live Cell Caspase 3/7 Imaging Kit *Green Fluorescence*
Leverandør: AAT BIOQUEST
20130
AATB20130EA 5150 NOK
AATB20130
Cell Meter™ Live Cell Caspase 3/7 Imaging Kit *Green Fluorescence*
Assay Cellulær analyse
Cell Meter™ Live Cell No Wash Caspase 3/7 Imaging Kit uses our recently developed cell-permeable fluorogenic caspase substrate, ApoSight™ Green Caspase 3/7, the first fluorogenic probe, for the direct detection of caspase activities in live cells.
  • Based on FMK inhibitor
  • Direct caspase detection
  • Suitable for all major fluorescence platforms
NB: For research use only.