2’,3’-cGAMP maleimide
Leverandør: AAT BIOQUEST
20336
AATB20336EA 15575 NOK
AATB20336
2’,3’-cGAMP maleimide
2’,3’-cGAMP maleimide
2’,3’-cGAMP maleimide is an excellent reagent to covalently conjugate 2’,3’-cGAMP to thiol containing-or thiol-modified biological targets, such as proteins, peptides, and nucleic acids.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only