Cal-500™ AM

Leverandør: AAT BIOQUEST
Warning

20412
AATB20412EA 3600 NOK
AATB20412
Cal-500™ AM
Cal-500™ AM

Calcium measurement is critical for numerous biological investigations.


  • Higher sensitivity
  • Specific to intracellular calcium
  • Steady signal
  • Cell permeant


NB: For research use only.

MW: 1019.92 g/mol
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR