Cal Green™ 1 hexapotassium salt
Leverandør: AAT BIOQUEST
Warning
20500
AATB20500EA 2700 NOK
AATB20500
Cal Green™ 1 hexapotassium salt
Cal Green™ 1 hexapotassium salt
Cal Green™ 1 (AAT Bioquest) is the same molecule to Calcium Green-1 (Invitrogen).
  • Calcium-dependent fluorescence
  • Steady signal
  • Cell impermeant
NB: For research use only.
ADR: 5,II