Cal Green™ 1, AM calcium indicator

Leverandør: AAT BIOQUEST
Warning

20501 20502
AATB20501EA 2700 NOK
AATB20501 AATB20502
Cal Green™ 1, AM calcium indicator
Calcium Green 1AM

Cal Green™ 1 (AAT Bioquest) is the same molecule to Calcium Green-1 (Invitrogen).


  • Cell permeable
  • Calcium-dependent fluorescence

Formel: C₅₉H₅₃Cl₂N₃O₂₆
CAS-nummer: 186501-28-0
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=accf2d02e2f0f350e532bc4781c9386557b1e467&vid=e4d9b52904aab8cb09c5bff4b7eec4380abf54e5&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR