OG488 BAPTA-1 AM Cell permeant [equivalent to Oregon Green® 488 BAPTA-1, AM]

Leverandør: AAT BIOQUEST
Warning

20507
AATB20507EA 3600 NOK
AATB20507
OG488 BAPTA-1 AM Cell permeant [equivalent to Oregon Green® 488 BAPTA-1, AM]
OG488 BAPTA-1 AM

OG488 BAPTA -1 AM is the same molecule of Oregon Green 488 BAPTA-1 AM ester.


  • Calcium-dependent fluorescence
  • Steady signal
  • Cell permeant


NB: For research use only.

Formel: C59H53F2N3O26
MW: 1258.07 g/mol
CAS-nummer: 244167-57-5
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR