BAPTA-AM ≥95%, Ultra Pure Grade

Leverandør: AAT BIOQUEST
Warning

Synonymer: Tetrakis(acetoxymethyl)-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N,N-tetraacetate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) Ester, BAPTA-AM ester

21002
AATB21002EA 1800 NOK
AATB21002
BAPTA-AM ≥95%, Ultra Pure Grade
BAPTA-AM

BAPTA AM is cell-permeable version of BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid), a cell-permeable calcium chelator.

Formel: C₃₄H₄₀N₂O₁₈
MW: 764.69 g/mol
MDL nummer: MFCD00036696
CAS-nummer: 126150-97-8
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR