EGTA, AM 10 mM calcium chelator

Leverandør: AAT BIOQUEST
Warning

21006
AATB21006EA 2700 NOK
AATB21006
EGTA, AM 10 mM calcium chelator
EGTA, AM

EGTA AM is cell-permeable version of EGTA (ethylene glycol tetraacetic acid), a cell-permeable calcium chelator.


  • High selectivity
  • Cell permeable

Formel: C₂₆H₄₀N₂O₁₈
CAS-nummer: 99590-86-0
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da83144da75fbc1b56b44de4405fec9eeadbb881&vid=bfeae64a18f09426198fa5402570e8137b2bec53&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR