FLUO 3/AM

Leverandør: AAT BIOQUEST
Warning

Synonymer: 4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, 1-[Amino-5-(2,7-dichloro-6-acetomethoxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid pentaacetoxy-methyl ester

21012
AATB21012EA 71625 NOK
AATB21012
FLUO 3/AM
FLUO 3/AM

Calcium measurement is critical for numerous biological investigations.

Formel: C₅₁H₅₀Cl₂N₂O₂₃
MW: 1129.85 g/mol
CAS-nummer: 121714-22-5
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR