Fluo-3, pentaammonium salt calcium indicator

Leverandør: AAT BIOQUEST
Warning

Synonymer: Fluo-3, 1-[2-Amino-5-(2,7-dichloro-6-hydroxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid .5NH3

21018
AATB21018EA 2700 NOK
AATB21018
Fluo-3, pentaammonium salt calcium indicator
Fluo-3, pentaammonium salt

Calcium measurement is critical for numerous biological investigations.


  • Intracellular probe
  • Calcium-dependent fluorescence

MW: 854,69 g/mol
CAS-nummer: 339221-91-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41a8b248de88f9c3a72bbda7081cf1970388ebe9&vid=59fa0058ce10f098a0ff8e7cf5b62ec1bda153c6&prid=web&clid=SAPDAM
New Product

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR