Fura-FF, AM 95

Leverandør: AAT BIOQUEST
Warning

Synonymer: Fura-2FF, AM

21027
AATB21027EA 3600 NOK
AATB21027
Fura-FF, AM 95
Fura-FF, AM

Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.

Formel: C₄₃H₄₃F₂N₃O₂₄
MW: 1023.8 g/mol
CAS-nummer: 348079-12-9
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR