Rhod-2 AM (Bromide) ≥95%, Ultra Pure Grade

Leverandør: AAT BIOQUEST
Warning

21062 21063 21064
AATB21062EA 3600 NOK
AATB21062 AATB21063 AATB21064
Rhod-2 AM (Bromide) ≥95%, Ultra Pure Grade
Rhod-2 AM (Bromide)

Calcium measurement is critical for numerous biological investigations.

Formel: C₅₂H₅₉BrN₄O₁₉
CAS-nummer: 145037-81-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7f4d1585f24a2cb85827e15f8b165cc3b93fb2a&vid=410926bfc7570104b03e13ee875ede0887fd13aa&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR