Rhod-5N AM

Leverandør: AAT BIOQUEST
Warning

21070
AATB21070EA 4500 NOK
AATB21070
Rhod-5N AM
Rhod-5N AM

Calcium measurement is critical for numerous biological investigations.


  • Calcium-dependent fluorescence
  • Low binding affinity
  • Cell permeant


NB: For research use only.

MW: 1154.92 g/mol
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR