BCECF AM ester, Ultra Pure Grade calcium indicator

Leverandør: AAT BIOQUEST
Warning

Synonymer: BCECF AM

21203
AATB21203EA 4500 NOK
AATB21203
BCECF AM ester, Ultra Pure Grade calcium indicator
BCECF AM ester

Intracellular pH plays an important modulating role in many cellular events, including cell growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, cell adhesion and other cellular processes.


  • Excellent intracellular pH measurement
  • Ultrapure grade

Formel: C₄₂H₄₀O₂₁
MDL nummer: MFCD00036969
CAS-nummer: 117464-70-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42713cce4e4173f3fad32ad83630bc0a0bc36893&vid=551a7ca767dc2cbfd9b78c943dc74e6e649c42bc&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR