Protonex™ Green 500-PEG12 SE
Leverandør: AAT BIOQUEST
21219
AATB21219EA 3175 NOK
AATB21219
Protonex™ Green 500-PEG12 SE
Protonex™ Green 500-PEG12 SE
The amino-reactive Protonex™ Green 500 SE (#21216) was developed for preparing Protonex™ Green 500 conjugates as fluorescent pH probes.
  • Stable for 24 months
Quality tested in-house
NB: For Research Use Only