5-(and-6)-Carboxy SNARF-1 AM ester
Leverandør: AAT BIOQUEST
21229
AATB21229EA 6300 NOK
AATB21229
5-(and-6)-Carboxy SNARF-1 AM ester
5-(and-6)-Carboxy SNARF-1 AM ester
5-(and-6)-Carboxy SNARF-1 is an excellent pH probe for monitoring pH changes in the range of 6.5 and 8.5.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only