Protonex™ Red 600 PEG12 maleimide
Leverandør: AAT BIOQUEST
21239
AATB21239EA 5450 NOK
AATB21239
Protonex™ Red 600 PEG12 maleimide
Protonex™ Red 600 PEG12 maleimide
Protonex™ Red 600 PEG12 maleimide is a thiol-reactive, pH-sensitive dye that has been specifically engineered for use in developing fluorescent pH probes applicable to live cell assays focusing on processes such as antibody internalization, endocytosis, and phagocytosis.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only