5-(and-6)-Carboxy SNARF-5F AM ester
Leverandør: AAT BIOQUEST
21248
AATB21248EA 6300 NOK
AATB21248
5-(and-6)-Carboxy SNARF-5F AM ester
5-(and-6)-Carboxy SNARF-5F AM ester
5-(and-6)-Carboxy SNARF-5F AM Ester is an excellent cell-permeable pH probes for ratiometrically monitoring pH changes in live cells.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only