Zinquin

Leverandør: AAT BIOQUEST
Warning

21260
AATB21260EA 1800 NOK
AATB21260
Zinquin
Zinquin

Zinquin is a zinc-sensitive, cell-impermeable fluorescent probe.

Formel: C₁₉H₁₈N₂O₅S
MDL nummer: MFCD03452711
CAS-nummer: 151606-29-0
ADR: 5,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR