Zinquin AM, Ultra Pure Grade
Leverandør: AAT BIOQUEST
21261
AATB21261EA 3600 NOK
AATB21261
Zinquin AM, Ultra Pure Grade
Zinquin AM
Zinquin AM ester is a lipophilic, zinc-sensitive, cell-permeable fluorescent probe.
  • Cell permeable
  • Zinc-dependent fluorescence
  • Multiple applications
NB: For research use only.