RH795 ≥95%

Leverandør: AAT BIOQUEST
Warning

21495
AATB21495EA 2300 NOK
AATB21495
RH795 ≥95%
RH795

RH 795 is a neuron-tracing dye.

Formel: C₂₆H₃₉Br₂N₃O₂
MW: 585.42 g/mol
CAS-nummer: 172807-13-5
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR