Native Dextran (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Leverandør: AAT BIOQUEST
21700
AATB21700EA 2700 NOK
AATB21700
Native Dextran (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Proteiner
FITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.
  • Well-retained in cells
  • 20000 mw
  • Multiple applications
NB: For research use only.
Bestill nå
SPESIFIKASJONER
Protein/peptide name Dextran
Protein synonyms Manninotriose|Macrodex
Environmentally Preferable
Protein/peptide type Native
Arter
Kilde Eukaryotic cells
Konjugering FITC (Fluorescein Isothiocyanate)