Violet 660 sodium salt, CytoCalcein™ excited at 405 nm
Leverandør: AAT BIOQUEST
21903
AATB21903EA 3175 NOK
AATB21903
Violet 660 sodium salt, CytoCalcein™ excited at 405 nm
Violet 660 sodium salt
CytoCalcein™ Violet 660 is the esterase-cleaved product of CytoCalcein™ Violet 660 AM in live cells.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only