Violet 660 AM, CytoCalcein™ excited at 405 nm
Leverandør: AAT BIOQUEST
21904
AATB21904EA 3175 NOK
AATB21904
Violet 660 AM, CytoCalcein™ excited at 405 nm
Violet 660 AM
CytoCalcein™ Violet 660 AM ester is designed for labeling live cells in the same way as Calcein, AM does.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only