Calcein AM 90

Leverandør: AAT BIOQUEST
Warning

Synonymer: Calcein-AM, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester, Calcein acetoxymethyl ester, Tetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate

22002
AATB22002EA 1800 NOK
AATB22002
Calcein AM 90
Calcein AM

Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein.

Formel: C₄₆H₄₆N₂O₂₃
MW: 994.86 g/mol
MDL nummer: MFCD05861516
CAS-nummer: 148504-34-1
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR