Tetracysteine FLASH Reagent *2 mM DMSO Solution*
Leverandør: AAT BIOQUEST
22330
AATB22330EA 6650 NOK
AATB22330
Tetracysteine FLASH Reagent *2 mM DMSO Solution*
Assay Proteinanalyser
FLASH is a fluorescein derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Fluorescein derivative
  • High affinity for thiols
  • Multiple applications
NB: For research use only.