Tetracysteine REASH Reagent *2 mM DMSO Solution*
Leverandør: AAT BIOQUEST
22332
AATB22332EA 6650 NOK
AATB22332
Tetracysteine REASH Reagent *2 mM DMSO Solution*
Assay Proteinanalyser
REASH is a resorufin derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Resorufin derivative
  • High affinity for thiols
  • Multiple applications
NB: For research use only.