Tetracysteine REASH Acetate *2 mM DMSO Solution*
Leverandør: AAT BIOQUEST
22336
AATB22336EA 6650 NOK
AATB22336
Tetracysteine REASH Acetate *2 mM DMSO Solution*
Assay Proteinanalyser
REASH acetate is a cell-permeable version of REASH, which is a resorufin derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Fluorescein derivative
  • High affinity for thiols
  • Multiple applications
NB: For research use only.