MycoLight™ Fluorescence Live/Dead Bacterial Imaging Kit

Leverandør: AAT BIOQUEST

22411
AATB22411EA 7200 NOK
AATB22411
MycoLight™ Fluorescence Live/Dead Bacterial Imaging Kit
Assay Immunologisk analyser

AAT Bioquest's MycoLight™ Fluorescence Live/Dead Bacterial Imaging Kit provides two-colour fluorescence assay for visualizing live and dead bacteria through fluorescent microscope.


  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for fluorescent microscopy


NB: For research use only.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR