Live or Dead™ Cell Viability Assay Kit *Red/Blue Dual Fluorescence*

Leverandør: AAT BIOQUEST

22788
AATB22788EA 5400 NOK
AATB22788
Live or Dead™ Cell Viability Assay Kit *Red/Blue Dual Fluorescence*
Assay Cellulær analyse

This Live or Dead™ Cell Viability Assay Kit uses two fluorescent indicators: Cellbrite™ Red (Ex/Em = 613/631 nm) for labelling viable cells and a cell-impermeable DNA-binding dye Nuclear Blue™ DCS1 (Ex/Em = 360/450 nm) for labelling dead cells with damaged membranes.


  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
  • Results in under 2 hours


NB: For research use only.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR