Cell Meter™ Caspase 3/7 Activity Apoptosis Assay Kit *Green Fluorescence*

Leverandør: AAT BIOQUEST

22796
AATB22796EA 3600 NOK
AATB22796
Cell Meter™ Caspase 3/7 Activity Apoptosis Assay Kit *Green Fluorescence*
Assay Cellulær analyse

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.


  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms


NB: For research use only.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR