Native Amanita phalloides Phalloidin (from Eukaryotic cells), Texas Red®
Leverandør: AAT BIOQUEST
23103
AATB23103EA 2700 NOK
AATB23103
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Texas Red®
Proteiner
This red fluorescent phalloidin conjugate (equivalent to Texas Red®-labeled phalloindin) selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
NB: For research use only.