Phalloidin, XFD350
Leverandør: AAT BIOQUEST
23150
AATB23150EA 5400 NOK
AATB23150
Phalloidin, XFD350
Proteiner
XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).
  • Well-cited conjugate
  • High affinity and selectivity
  • In house quality control
NB: For research use only.