Cell Meter™ 2-NBDG Glucose Uptake Assay Kit

Leverandør: AAT BIOQUEST

23500
AATB23500EA 9000 NOK
AATB23500
Cell Meter™ 2-NBDG Glucose Uptake Assay Kit
Assay Cellulær analyse

Glucose metabolism, a process which converts glucose into energy, is a primary source of energy supply in most organisms.


  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms


NB: For research use only.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR