MycoLight™ vPCR350
Leverandør: AAT BIOQUEST
24210
AATB24210EA 2325 NOK
AATB24210
MycoLight™ vPCR350
MycoLight™ vPCR350
MycoLight™ vPCR350 is a novel non-fluorescent DNA modifier specifically designed for viability PCR (vPCR) targeting microorganisms such as bacteria, eukaryotes, viruses, and fungi.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only