AzoDye-3 carboxylic acid
Leverandør: AAT BIOQUEST
2470
AATB2470EA 4925 NOK
AATB2470
AzoDye-3 carboxylic acid
AzoDye-3 carboxylic acid
AzoDye-3 Carboxylic Acid is an excellent building block for labeling amino-containing molecules.
  • Stable for 12 months
Quality tested in-house
NB: Research use only (RUO).