mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
Leverandør: AAT BIOQUEST
25450
AATB25450EA 2375 NOK
AATB25450
mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
Systemer for proteinkobling og -merking
mFluor™ Violet 450 labelled WGA is well-excited by the violet laser, emitting a bright blue fluorescence at 445 nm.
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
NB: For research use only.